Distillation apparatus, flat flange, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
B280-09 B280-06 B280-03 B280-12 B280-15
BOHLB280-09EA 9510 EUR
BOHLB280-09 BOHLB280-06 BOHLB280-03 BOHLB280-12 BOHLB280-15
Distillation apparatus, flat flange, BOLA
Onderdelen voor destillatie Destillatiesets
PTFE/PFA.
  • Temperature resistance: from −200 to +200 °C
  • Universal chemical resistance
  • Vacuum: suitable
Suitable for the distillation of strong alkaline or acid products as well as very aggressive solvents.