Magnetic stirring bars, coloured PTFE-encapsulated, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
C368-28 C368-60 C368-56 C368-52 C368-48 C368-40 C368-32 C368-20 C368-16 C368-12 C368-08 C368-36
BOHLC368-28EA 4.3 EUR
BOHLC368-28 BOHLC368-60 BOHLC368-56 BOHLC368-52 BOHLC368-48 BOHLC368-40 BOHLC368-32 BOHLC368-20 BOHLC368-16 BOHLC368-12 BOHLC368-08 BOHLC368-36
Magnetic stirring bars, coloured PTFE-encapsulated, BOLA
Roerders Magnetische roerstaven
Magnetic stirrer bars with coloured PTFE-encapsulated magnetic core (Alnico 5), conically tapered ends.
  • Chemical resistant
  • Coloured to provide better distinction
Temperature resistance: −200 to +250 °C.