Stirrer blades for centrifugal stirrer shafts, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
C407-04 C407-06 C407-08
BOHLC407-04EA 13.2 EUR
BOHLC407-04 BOHLC407-06 BOHLC407-08
Stirrer blades for centrifugal stirrer shafts, BOLA
Roerders Roerbladen
Centrifugal stirrer blades made of PTFE consisting of 2 paddles, bolt and clamp ring.
  • Resistant to chemicals
  • Temperature resistance from –200 to +250 °C
Suitable for vessels with a 24/29 ground joint and centrifugal stirrer shafts.