Stirrer Bearings, glass
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
C425-09 C425-12 C425-08 C425-14 C428-12 C428-08 C425-06
BOHLC425-09EA 123 EUR
BOHLC425-09 BOHLC425-12 BOHLC425-08 BOHLC425-14 BOHLC428-12 BOHLC428-08 BOHLC425-06
Stirrer Bearings, glass
Roerders Roergereedschap
Combination of borosilicate glass piece with ground joint, an interior PTFE shaft guide with integrated special gasket and a GL screw cap made
of polyphenylene sulphide.
  • Suitable for vacuum
  • Perfect bearing for stirrer shafts made of stainless steel, glass and for BOLA stirrer shafts
  • Temperature resistance: –15 to +200 °C
  • Chemical resistance
The special gasket made of PTFE and an FPM o-ring which is compressed by a GL screw cap provide a good sealing of the stirrer shaft.
This gasket can be exchanged after wearing.
Certificaten: FDA conform