Slip-on baffle, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
C490-12 C490-10
BOHLC490-12EA 25.6 EUR
BOHLC490-12 BOHLC490-10
Slip-on baffle, BOLA
Roerders Roergereedschap
PTFE, with PFA support ring. The baffle can be mounted at any position on a temperature probe or a solo stirrer shaft.
  • Vacuum suitable
  • Universal chemical resistance
  • Temperature resistant from −200 to +250 °C
Certificaten: DIN 28131