Tubing connectors, cross-shaped, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
D573-08
BOHLD573-08EA 68 EUR
BOHLD573-08
Tubing connectors, cross-shaped, BOLA
Fittingen en connectoren
The Cross-shaped fitting, made of PTFE with four connectors for elastic tubing.
  • Universal chemical resistance
  • Temperature resistance from -200 up to 250 °C
  • Vacuum: suitable