Adapter, standard ground joint on GL thread, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
D734-50 D734-54
BOHLD734-50EA 177 EUR
BOHLD734-50 BOHLD734-54
Adapter, standard ground joint on GL thread, BOLA
Adapters Adapters met NS-verbindingen
PP ( polypropylene) screw cap, blue with joint insert made from PTFE.
  • Excellent chemical resistance
  • Temperature resistant from -50 to +250 °C
  • Suitable for vacuum applications
For installing components with standard ground joints (coolers, mixing closures and more) on glass bottles or GL threaded connectors. Screw cap with moving joint insert. The distributor head can be moved independently of the screw cap. The entire structure can be placed on another vessel easily and without the need for prior disassembly, avoiding the risk of kinking or twisting already assembled tubes.