Sample dippers, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
H1138-24 H1138-16 H1138-08
BOHLH1138-24EA 376 EUR
BOHLH1138-24 BOHLH1138-16 BOHLH1138-08
Sample dippers, BOLA
Monsternemers Monsterdompelaars
Sample vessels suitable for washing, rinsing or dipping solids in aggressive, or pure substances.
  • Chemical resistance and temperature resistance
  • Handle and holes in wall and botton
Certificaten: FDA conform.