O-rings for laboratory flat flanges, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
H969-18 H969-55 H969-45 H969-25 H969-75
BOHLH969-18EA 22.7 EUR
BOHLH969-18 BOHLH969-55 BOHLH969-45 BOHLH969-25 BOHLH969-75
O-rings for laboratory flat flanges, BOLA
Afdichtingen Afdichtringen
FEP. Sealing for flat flange with groove.
  • Temperature resistance: from –60 to +205 °C
  • Excellent chemical resistance
Silicone core with seamless FEP coating; manufactured according to DIN 12214;1996-12; flexible, almost universal chemical resistance.