Screw caps for bottles with GL threads, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
H986-04 H986-02 H986-03 H986-05 H986-01
BOHLH986-04EA 53.6 EUR
BOHLH986-04 BOHLH986-02 BOHLH986-03 BOHLH986-05 BOHLH986-01
Screw caps for bottles with GL threads, BOLA
Sluitingen Flesdoppen
Screw cap with handy knurl, made of PTFE.
  • Very good chemical resistance
  • Temperature resistant from −200 to +250 °C
Suitable for bottles and vessels with GL threads.