b.safe Blind Fitting PFAS Analytic
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
M401-01
BOHLM401-01EA 53.5 EUR
BOHLM401-01
b.safe Blind Fitting PFAS Analytic
Fittingen en connectoren
To close unused connections on b.safe Caps PFAS Analytic.
  • Made of PE
  • For PFAS analysis
  • With ergonomically shaped grip surface
  • easy mounting
Blind Fittings made of PE for tight closure of unused connectors on b.safe Caps PFAS Analytic. With ergonomic gripping surface for easy assembly.