b.safe Screw Caps
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
M575-01
BOHLM575-01EA 8.8 EUR
BOHLM575-01
b.safe Screw Caps
Sluitingen Flesdoppen
Screw cap made of PP with handy knurl and internal PTFE/silicone gasket. After assembly, the product is only exposed to PTFE.
  • Made of PP
  • Very good chemical resistance
  • Equipped with PTFE/silicone seal
Screw caps are designed for closing unused connections on a b.safe Distributor for flushing bottles, or also to close eluent bottles during the flushing process.