Extension cables for temperature probes, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
P1724-08 P1724-16 P1724-38 P1724-46
BOHLP1724-08EA 183 EUR
BOHLP1724-08 BOHLP1724-16 BOHLP1724-38 BOHLP1724-46
Extension cables for temperature probes, BOLA
Kabels
PTFE, 4-pole measuring cable and Lemo coupling. It is suitable for all BOLA temperature probes with coupling type Lemo, socket size 1.
  • Excellent chemical resistance
  • Temperature resistant from -200 to +250 °C