Sink XL, b.safe
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
R1250-40
BOHLR1250-40EA 1110 EUR
BOHLR1250-40
Sink XL, b.safe
Kranen en gootstenen Gootstenen
Made of static dissipative PE-EX, with sloped bottom for complete emptying.
  • Made of static dissipative PE-EX
  • With mounted grounding cable
  • Can be covered completely
To be integrated in laboratory hoods or working tables for an easy disposal of large quantities of liquid solvent waste. Thread connection GL 25 to connect b.safe waste tubes that transport liquid solvent wastes directly in canisters.
Drain Ø 60 mm with removable sieve. Bottom drain with male thread GL 25 for direct connection to flexible b.safe waste tubes. Grounding connection at theMore Product Information
Order Now
Specificaties
Environmentally Preferable