Grounding cables, b.safe
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
R601-01 R603-01 R603-03 R604-01 R605-01 R606-01 R612-01 R612-03 R615-03 R606-05 R607-03
590-0337EA 44.1 EUR
590-0337 590-0338 590-0339 590-0340 590-0341 590-0342 590-0343 590-0344 590-0345 BOHLR606-05 BOHLR607-03
Grounding cables, b.safe
Aardingen Aardingskabels
For earth connection of vessels and components made of static dissipative materials.
  • Choose the suitable connection to earth that we offer in our range of grounding cables with different connectors
  • Customised versions can be offered on request
  • Suitable grounding accessories for b.safe products
The scope of delivery of the ring cable lug includes screw, nut and washer.