Five-fold tap banks, Discofix® C
Leverancier: B. Braun
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
16609C
BRAU16609CEA 691 EUR
BRAU16609C
Five-fold tap banks, Discofix® C
Infusen
Five-fold tap banks are used for administration of medicines and medicines such as anesthetics, cytostatics, antibiotics, narcoleptics, immunosuppressants and lipids.
  • Leaks caused by stress cracks, contamination and air embolism are prevented
  • Easy access through the rotary adapter for injections and aspirations
  • Control of the 45° position by 'clicking'
  • Pressure resistant up to 2 bar
  • PVC and DEHP-free
Order Now
Specificaties
Environmentally Preferable