Labels for BBP3x printers, type B-7569
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
120920 120921 120923 120928 120925 120926 120927 120929 120924 120922 120906 120910 120914 120918 120930 120901 120935 120919 120907 120911 120915 120903 120933 120905 120900 120902 120904 120908 120909 120912 120913 120916 120917 120931 120932 120934
BRDY120920EA 71.13 EUR
BRDY120920 BRDY120921 BRDY120923 BRDY120928 BRDY120925 BRDY120926 BRDY120927 BRDY120929 BRDY120924 BRDY120922 BRDY120906 BRDY120910 BRDY120914 BRDY120918 BRDY120930 BRDY120901 BRDY120935 BRDY120919 BRDY120907 BRDY120911 BRDY120915 BRDY120903 BRDY120933 BRDY120905 BRDY120900 BRDY120902 BRDY120904 BRDY120908 BRDY120909 BRDY120912 BRDY120913 BRDY120916 BRDY120917 BRDY120931 BRDY120932 BRDY120934
Labels for BBP3x printers, type B-7569
Etiketten
B-7569 vinyl is ideal for indoor/outdoor facility labels, including 5S labels, safety labels, warning labels, pipe markers, equipment labels and visual workplace labels.
  • Gloss finish
  • Up to three years outdoor use
  • Temperature range from –20 to +75 °C