Preprinted Tags "Energy Source, Control Panel"
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
138396 138397 138398 138732 138733 138734 138735 138736 138737 138738
BRDY138396EA 57.91 EUR
BRDY138396 BRDY138397 BRDY138398 BRDY138732 BRDY138733 BRDY138734 BRDY138735 BRDY138736 BRDY138737 BRDY138738
Preprinted Tags "Energy Source, Control Panel"
Etiketten
These energy source control panel tags are constructed of rigid PP with round corners.
  • Permanent acrylic adhesive
  • Helps the lockout process quicker and easier for higher safety
Used to mark your facility's points of control of control for energy sources.
Colour: Orange on white
Certificaten: ATEX
Verpakking: 25 tags per pack.