Preprinted Tags "Energy Source, Steam"
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
138423 138424 138425 138426 138427 138761 138762 138763 138764 138765
BRDY138423EA 57.91 EUR
BRDY138423 BRDY138424 BRDY138425 BRDY138426 BRDY138427 BRDY138761 BRDY138762 BRDY138763 BRDY138764 BRDY138765
Preprinted Tags "Energy Source, Steam"
Etiketten
These steam energy source tags are constructed of PP with round corners with one hole.
  • Permanent acrylic adhesive
  • Helps the lockout process quicker and easier for higher safety
Used to mark your facility's points of control of control for energy sources.
Colour: Black on orange
Certificaten: ATEX
Verpakking: 25 tags per pack.