Core Inkjet Polypropylene Roll Labels, 76,20 mm

Leverancier: Brady
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

153635 153636 153651 153652 153653 153654 153656
BRDY153635EA 200.71 EUR
BRDY153635 BRDY153636 BRDY153651 BRDY153652 BRDY153653 BRDY153654 BRDY153656
Core Inkjet Polypropylene Roll Labels, 76,20 mm
Etiketten
This B-7425J polypropylene material is ideal for full-colour identification and barcode applications where resistance to liquid nitrogen, autoclave conditions, and harsh chemicals is required.

  • White polypropylene with acrylic adhesive is inkjet printable
  • Perfect for laboratory identification where labels may come in contact with harsh chemicals
  • Designed for roll inkjet printers
  • Service temperature of −80 to +70 °C (−112 to +158 °F)
  • Matte finish

Applications include barcode labelling, cold temperature labelling, COVID-19, general identification, hazardMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR