Self-Laminating Cryogenic Polyester Labels for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
170882 170877 170767 170765 170875 170766 170768 170769 170878 170879 170880 170881 170883
BRDY170882EA 71 EUR
BRDY170882 BRDY170877 BRDY170767 BRDY170765 BRDY170875 BRDY170766 BRDY170768 BRDY170769 BRDY170878 BRDY170879 BRDY170880 BRDY170881 BRDY170883
Self-Laminating Cryogenic Polyester Labels for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Etiketten
Securely label laboratory tubes and vials for cryogenic storage with this self-laminating, ultra-thin polyester label material (B-461). Print almost anywhere with a compact Brady portable label printer.
  • For use on smooth, curved surfaces
  • Withstands extreme temperatures from −196 to +130 °C
  • Chemical-resistant
  • Transparent overlaminate protects print
Versatile labels are ideal for curved surfaces such as vials, centrifuge tubes, test tubes, straws, and slides. Self-Laminating Cryogenic Polyester Labels are able to withstand extreme temperatures from −196More Product Information
Order Now
Specificaties
Environmentally Preferable