Printer Ribbons, M710 R4400 Series
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
173192
BRDY173192EA 76.22 EUR
BRDY173192
Printer Ribbons, M710 R4400 Series
Printerlinten
One-color ribbon for printing a single color onto a label.
  • R4400 series resin formulation offers excellent resistance to solvents, scratches, and high heat when printed on the appropriate Brady® label material
  • Recommended for printing on glossy or very smooth label stocks, such as B-483 Ultra aggressive polyester and B-423 permanent polyester
  • Self-contained M710 series ribbon cartridges insert straight into the printer with ease
M710 Series snap-in print ribbons enable 5 second ribbon supply changes. No calibrating, no threading, no waste, no hassles,More Product Information