Self-Laminating Self-Extinguishing Tedlar Wrap Around Label for M610, M611, BMP61, M710 (with Media Adapter) and BMP71 (with Media Adapter)
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
174226 174238 174281
BRDY174226EA 91.22 EUR
BRDY174226 BRDY174238 BRDY174281
Self-Laminating Self-Extinguishing Tedlar Wrap Around Label for M610, M611, BMP61, M710 (with Media Adapter) and BMP71 (with Media Adapter)
Etiketten
The B-642 is a thin tape with self-extinguishing properties that has excellent flexibility for wrapping on small diameter wires.
  • Ideal for use in aerospace, defense and mass transit industries
  • Polyvinylfluoride film provides good self-extinguishing properties and solvent resistance
  • For use in dot matrix and thermal transfer printers
  • Matte finish
Ideal for data and telecommunications labelling, electrical labelling, and wire and cable labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer andMore Product Information