Prohibition Signs
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
241035 197582 250035 250037
BRDY241035EA 3.28 EUR
BRDY241035 BRDY197582 BRDY250035 BRDY250037
Prohibition Signs
Pictogrammen
Brady B-7541 is a surface printed white polyester film with a permanent, acrylic based, pressure sensitive adhesive and over-laminated with a clear polyester film.
  • B-7541 is used for pipe markers and safety signs
  • B-7541 gives excellent adhesion to low surface energy surfaces, such as polypropylene and ABS, as well as on most powder coatings
  • Standard ink colors are black, red (RAL 3001-GL), yellow (RAL 1003-GL), blue (RAL5005-GL) and green (RAL 6032-GL)
Order Now
Specificaties
Environmentally Preferable