Self-laminating Polyester Labels for M710
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
311977 311989 311988 311975 311992 311973 311974 311976 311979
BRDY311977EA 63.01 EUR
BRDY311977 BRDY311989 BRDY311988 BRDY311975 BRDY311992 BRDY311973 BRDY311974 BRDY311976 BRDY311979
Self-laminating Polyester Labels for M710
Etiketten
Self-laminating polyester labels for M710 printer.
  • Self-laminating polyester has a permanent acrylic adhesive and is thermal transfer printable
  • Good print smudge resistance, solvent resistance, and high/low temperature performance
Ideal for laboratory labelling applications.