Linerless Polyester Cable Tags for M610 and M611
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
312077 312054 312079 312083 312078 312063 312064 312080 312055 312082 312053 312081 312052
BRDY312077EA 164.26 EUR
BRDY312077 BRDY312054 BRDY312079 BRDY312083 BRDY312078 BRDY312063 BRDY312064 BRDY312080 BRDY312055 BRDY312082 BRDY312053 BRDY312081 BRDY312052
Linerless Polyester Cable Tags for M610 and M611
Etiketten
The B-7598 polyester cable identification tag has been developed to keep cables identified in outdoor environments.
  • Gloss finish
The B-7598 polyester cable identification tags offer up to 10 year outdoor legibility and durability, and are available in seven colours. Applications include data and telecommunications labelling, and wire and cable labelling.
Certificaten: Tags supplied in white, black, or silver finish are Halogen-free per IEC 61249-2-21 (2003-11).
Verpakking: 100 tags per roll.