Polyester Tags for M710, M610 and M611, Bulk
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
312147 312168 312161 312154
BRDY312147EA 342.6 EUR
BRDY312147 BRDY312168 BRDY312161 BRDY312154
Polyester Tags for M710, M610 and M611, Bulk
Etiketten
The B-7598 polyester cable identification tag has been developed to keep cables identified in outdoor environments.
  • Gloss finish
The B-7598 polyester cable identification tag offers up to 10 year outdoor legibility and durability, and is available in seven colours. Applications include data and telecommunications labelling, and wire and cable labelling.
Certificaten: Tags supplied in white, black, or silver are Halogen-free per IEC 61249-2-21 (2003-11).
Order Now
Specificaties
Environmentally Preferable