Universal sorbent rolls
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
813828
BRDY813828EA 212.14 EUR
BRDY813828
Universal sorbent rolls
Absorptiematerialen en bescherming tegen morsen
These grey coloured medium weight, bonded and perforated universal sorbent rolls absorbs industrial liquids including oils, water, solvents, coolants.
  • Made of one-ply, meltblown, bonded
  • Strong and durable with a superior finish with less lint for improved abrasion resistance
Ideal to cover a large surface area for fast clean up of spills, leaks and drips. Suited for industrial applications where low cost and high absorbency are required.