Spill kit, spill truck
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
813888 813889
BRDY813888EA 2477.83 EUR
BRDY813888 BRDY813889
Spill kit, spill truck
Absorptiematerialen en bescherming tegen morsen
Spill Truck spill kits are ideal for larger spills.
  • Lid provides easy access to the products
  • Wheels allow excellent mobility indoors and outdoors
  • Weather and chemical resistant container protects sorbents from moisture, dirt and damage
Leveringsinformatie: Kit contains: 150 pads (400×500 mm), 36 socs (Ø 76×1220 mm), 20 socs (Ø 76×3660 mm), 16 pillows (430×480 mm), 50 wipes (300×380 mm), 1 pair of gloves, 1 pair of goggles and 30 disposal bags.