Bulb pipettes, BLAUBRAND® ETERNA, one mark, Class AS (one batch)
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2.30512 2.30507 2.30518
BRND2.30512EA 53.3 EUR
BRND2.30512 BRND2.30507 BRND2.30518
Bulb pipettes, BLAUBRAND® ETERNA, one mark, Class AS (one batch)
Pipetten Volpipetten
AR-Glas®.
  • Calibrated to deliver (TD, Ex)
  • Including one batch certificate
  • Ring mark and inscriptions in ETERNA amber stain
  • Total order from one batch: All supplied pipettes come from one batch for statistical testing equipment monitoring
Certificaten: DIN EN ISO 648; DE-M marking