Automatische buretten, Pellet-patroon, zonder tussenkraan, klasse B, Schellbach, SILBERBRAND
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
21911 21912 21913
BRND21911EA 200 EUR
BRND21911 BRND21912 BRND21913
Automatische buretten, Pellet-patroon, zonder tussenkraan, klasse B, Schellbach, SILBERBRAND
Buretten
Borosilicate glass 3.3, class B.
  • Calibrated to deliver 'TD, Ex'
  • Schellbach stripe, ring marks at major graduations
  • Automatic zeroing, needle-valve stopcock with PTFE spindle, without intermediate stopcock
Certificaten: DIN EN ISO 385