Sterilisation Boxes for Pipettes
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
28920 28925 28930 28935
BRND28920EA 58.5 EUR
BRND28920 BRND28925 BRND28930 BRND28935
Sterilisation Boxes for Pipettes
Dozen Pipetsterilisatiecontainers
These pipette boxes made of aluminum are ideal for the storage and sterilisation of pipettes with different lengths.
  • With inner silicone coating
  • Very good temperature resistance
  • Very good chemical resistance