Bulb pipettes, with individual certificate, BLAUBRAND®, two marks, Class AS
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
929734 929735 929738 929722 929723 929727 929721 929725 929732 929733
BRND929734EA 39.4 EUR
BRND929734 BRND929735 BRND929738 BRND929722 BRND929723 BRND929727 BRND929721 BRND929725 BRND929732 BRND929733
Bulb pipettes, with individual certificate, BLAUBRAND®, two marks, Class AS
Pipetten Volpipetten
AR-Glas®
  • Calibrated to deliver (TD, Ex)
  • Marks and inscriptions in high contrast blue enamel
  • Including an individual certificate
Certificaten: DIN EN ISO 648; DE-M marking