Volumetric flasks, class A, blue markings, BLAUBRAND®
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
937048 937045 937047
BRND937048EA 40.6 EUR
BRND937048 BRND937045 BRND937047
Volumetric flasks, class A, blue markings, BLAUBRAND®
Kolven Maatkolven
Borosilicate glass 3,3, with beaded rim.
  • Calibrated to contain (TC, In)
  • Marks and inscriptions in high contrast blue enamel
  • DE-M marking
* Wide neck
Certificaten: DIN EN ISO 1042
Bestelinformatie: Supplied with an individual certificate.