Bulb pipettes (one mark), BLAUBRAND®, Class AS, with DAkkS certificate
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
DAKKS29702 DAKKS29704 DAKKS29706 DAKKS29707 DAKKS29710 DAKKS29712 DAKKS29714 DAKKS29713 DAKKS29715 DAKKS29719 DAKKS29701 DAKKS29709 DAKKS29703 DAKKS29705 DAKKS29718
BRNDDAKKS29702EA 72.1 EUR
BRNDDAKKS29702 BRNDDAKKS29704 BRNDDAKKS29706 BRNDDAKKS29707 BRNDDAKKS29710 BRNDDAKKS29712 BRNDDAKKS29714 BRNDDAKKS29713 BRNDDAKKS29715 BRNDDAKKS29719 BRNDDAKKS29701 BRNDDAKKS29709 BRNDDAKKS29703 BRNDDAKKS29705 BRNDDAKKS29718
Bulb pipettes (one mark), BLAUBRAND®, Class AS, with DAkkS certificate
Pipetten Volpipetten
AR-Glas®.
  • Calibrated to deliver (TD, Ex)
  • Ring mark and inscriptions in blue enamel
  • Waiting time 5 seconds
  • With DAkkS calibration certificate
Certificaten: DIN EN ISO 648; DE-M marking.