Bottle top dispensers, Dispensette® S, DAkkS
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
DAKKS4600170 DAKKS4600171 DAKKS4600331 DAKKS4600341 DAKKS4600360 DAKKS4600361 DAKKS4600350 DAKKS4600351
BRNDDAKKS4600170EA 813 EUR
BRNDDAKKS4600170 BRNDDAKKS4600171 BRNDDAKKS4600331 BRNDDAKKS4600341 BRNDDAKKS4600360 BRNDDAKKS4600361 BRNDDAKKS4600350 BRNDDAKKS4600351
Bottle top dispensers, Dispensette® S, DAkkS
Flesdispensers
Fixed volume or variable volume, analogue-adjustable or Digital Easy Calibration.
  • With or without recirculation valve
  • Three different models of volume adjustment available: Fixed (Fixed-volume for standard applications; simple calibration adjustment with supplied tool) and Analog (Analog volume display, fast volume adjustment; simple calibration adjustment with supplied tool)
  • Digital EASY CALIBRATION: Easy to read digital display; dispensing volume can be set accurately and reproducibly. Easy Calibration technique for adjustment in seconds without tools;
More Product Information