Fittings, Uniflex™
Leverancier: BROEN-LAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
25330591036-09 25330111036-09
BROE25330591036-09EA 2260 EUR
BROE25330591036-09 BROE25330111036-09
Fittings, Uniflex™
Kranen en gootstenen Kranen
UniFlex™ BASIC fittings for special water are designed to secure maximum purity of the water in laboratory. The operation and configuration of the fittings are highly adapted to the requirements of a modern laboratory. These fittings are surface treated with polycoat powder coating, which is highly resistant to most chemicals, UV light and heat. Body made from LLDPE, POM, PP, PVDF / EPDM rubber.
  • Safe and simple installation
  • Ideal for corrosive media
  • Eliminate corrosion problems in connection with use of purified water
  • Spout with full swivel action
  • With
More Product Information