Anti-RAGE Rabbit Polyclonal Antibody

Leverancier: Boster Bio
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PB9469
BSBTPB9469EA 552 EUR
BSBTPB9469
Anti-RAGE Rabbit Polyclonal Antibody
Antilichamen
Rabbit IgG polyclonal antibody for Advanced glycosylation end product-specific receptor (AGER) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Type: Primary
Antigen: RAGE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR