Anti-NEST Chicken Polyclonal Antibody
Leverancier: Biosensis
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
C-1812-50
BSENC-1812-50EA 312 EUR
BSENC-1812-50
Anti-NEST Chicken Polyclonal Antibody
Antilichamen
Required for brain and eye development. Promotes the disassembly of phosphorylated vimentin intermediate filaments (IF) during mitosis and may play a role in the trafficking and distribution of IF proteins and other cellular factors to daughter cells during progenitor cell division. Required for survival, renewal and mitogen-stimulated proliferation of neural progenitor cells (By similarity). Ref: uniprot.org
Western blotting (1:1,000-1:5,000) and Immunocytochemistry (1:2,000-1:5,000). Biosensis recommends optimal dilutions/concentrations should be determined by the end user.