Anti-PRPH Mouse Monoclonal Antibody [clone: 7C5]

Leverancier: Biosensis
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

M-1400-100
BSENM-1400-100EA 312 EUR
BSENM-1400-100
Anti-PRPH Mouse Monoclonal Antibody [clone: 7C5]
Antilichamen
Peripherin is a class-III neuronal intermediate filament protein found in certain classes of neuron, most of which are located in the peripheral nervous system.

Application information: Western Blotting (WB), Immunocytochemistry (IC) and Immunohistochemistry (IH). A concentration of 0.5 - 2 ug/mL is recommended for WB. A concentration of 1-5 ug/mL is recommended for IC and IH. This antibody performs well on aldehyde fixed tissues. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

"Type:
Antigen:
Clonality:
Clone:
Conjugation:
Epitope:
Host:
Isotype:
Reactivity: "
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR