Anti-HA-Tag Rabbit Polyclonal Antibody

Leverancier: Biosensis
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

R-1307-100
BSENR-1307-100EA 455 EUR
BSENR-1307-100
Anti-HA-Tag Rabbit Polyclonal Antibody
Antilichamen
The Human influenza hemagglutin (HA) tag corresponds to a region (98-106 amino acids) from the HA molecule.

Antibody detects HA-tagged proteins (on amino- and carboxy termini) in transfected mammalian cells.
Application Information:
Western Blotting at a suggested dilution 1:500~1:1000. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Tag
Antigen: HA-Tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: HA epitope
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR