Petri dish platinum electrodes kit
Leverancier: BTX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
45-0500 45-0490 45-0491 45-0492 45-0501 45-0502
BTXI45-0500EA 5370 EUR
BTXI45-0500 BTXI45-0490 BTXI45-0491 BTXI45-0492 BTXI45-0501 BTXI45-0502
Petri dish platinum electrodes kit
Elektroden Elektroden voor elektroporatie
This electrode is designed for delicate and/or difficult tissue transfection. Ex vivo electropoartion is an efficient, effective method to introduce genes, drugs or any number of molecules into a tissue. Sandwich configuration ensures a homogeneous field of energy for optimum transfection.
  • The electrode is compromised of two parts, the petri dish and wand
  • The petri dish contains a platinum electrode chamber to secure the tissue
  • The wand incorporates an identical shaped platinum electrode, placed over the chamber to complete electroporation