Dropping bottles, LDPE, LaboPlast®
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0306-0030 0306-0100 0306-0050 0306-0500 0306-0010 0306-1000 0306-0250
BURK0306-0030EA 0.9 EUR
BURK0306-0030 BURK0306-0100 215-4290 BURK0306-0500 BURK0306-0010 BURK0306-1000 BURK0306-0250
Dropping bottles, LDPE, LaboPlast®
Flessen Druppelflessen
LDPE, translucent.
  • Cut off dropper tip before first use
  • Can be reclosed using the captive cap provided
  • Captive cap and dropper cap PE
Dropping bottles are particularly suitable for liquids which must be dispensed in small amounts, drop by drop or as a fine jet.