Dropping bottles, with drop dispenser and screw cap
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0307-0010 0307-0025 0307-0050
BURK0307-0010EA 1 EUR
BURK0307-0010 BURK0307-0025 BURK0307-0050
Dropping bottles, with drop dispenser and screw cap
Flessen Druppelflessen
LDPE, translucent.
  • Cut off dropper tip before first use
  • Reclosable
  • Precise drop dosing
Bottle with drop dispenser and separate screw cap.