Wash bottle, LaboPlast®
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0310-0010 0310-0025 0310-0050 0310-0100
BURK0310-0010EA 2.1 EUR
BURK0310-0010 BURK0310-0025 BURK0310-0050 BURK0310-0100
Wash bottle, LaboPlast®
Flessen Spuitflessen
LDPE, translucent.
  • Immersion tube can be pulled up
  • Good chemical resistance
  • Good temperature resistance
The swan-neck jet produces a fine, straight spray with no lateral spatter. Dangerous lateral spatter does not occur.
Order Now
Specificaties
Environmentally Preferable