Buckets, Round, HDPE
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2306-0015 2306-0010
BURK2306-0015EA 58.5 EUR
BURK2306-0015 BURK2306-0010
Buckets, Round, HDPE
Emmers
PE, white.
  • With contents scale, graduations 1000 ml
  • With spout
  • Handle made of metal
Laboratory bucket for transport, decanting and mixing. Safe emtying with wide spout.
Order Now
Specificaties
Environmentally Preferable