Emmers, rond, HDPE
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2307-0017 2307-0010 2307-0011 2307-0018
BURK2307-0017EA 28.9 EUR
BURK2307-0017 BURK2307-0010 BURK2307-0011 BURK2307-0018
Emmers, rond, HDPE
Emmers
HDPE, white or black.
  • With litre scale, graduation 1000 ml
  • With spout
  • White buckets suitable for use with foodstuffs
  • Black buckets not suitable for food
For transporting, decanting and mixing. Wide spout gives rapid but safe emptying. With plastic handle.