Transport- en opslagcontainers
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3414-0117 3414-0111 3414-0164 3414-0138 3414-0148 3414-0120 3414-0181 3414-0123
216-1723EA 28.7 EUR
216-1723 216-1724 BURK3414-0164 BURK3414-0138 BURK3414-0148 BURK3414-0120 BURK3414-0181 BURK3414-0123
Transport- en opslagcontainers
Dozen Transportdozen
PP, grey.
  • Temperature resistant from −40 to +90 °C
  • According to Euro standards
  • Lid and transport rollers available as accessory
  • Suitable for use with foodstuffs
Stable storage containers with external reinforcement ribs.