Meng- en doseercontainers
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3460-0200 3460-0100 3460-0060 3460-0500
216-1721EA 527 EUR
216-1721 BURK3460-0100 BURK3460-0060 BURK3460-0500
Meng- en doseercontainers
Flessen Voorraadflessen
The containers close completely tight. They have thick sides, they are seamless, impact-resistant and stress-free sintered.
  • LDPE
  • With 3/4" inner thread at the container base
  • Suitable for food
The wide-necked opening enables safe filling.