Forceps, stainless steel
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5386-0104 5386-0304 5386-0102
BURK5386-0104EA 4.7 EUR
BURK5386-0104 BURK5386-0304 BURK5386-0102
Forceps, stainless steel
Pincetten
These stainless steel forceps are suitable for precise laboratory tasks. The laboratory tweezers are made of stainless steel and feature a fluting at the ends and grip for secure handling.
  • Stainless steel (1.4301)
  • Flutings at the end and grip for secure handling
These laboratory forceps are suitable for precise work in the fields of chemistry, pharmacy, cosmetics and food and beverage.
Order Now
Specificaties
Environmentally Preferable